Assay Details
| Gene Symbol: gabrz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114742.2 | NP_001108214.2 | 5 - 6 | 653 | 73 | |
| GenBank mRNA | BC159112.1 | - | 5 - 6 | 653 | 73 | |
More Information
Set Membership: |
|

gabrz
Genomic Map