Assay Details
| Gene Symbol: hdhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328522.1 | NP_001315451.1 | 1 - 2 | 373 | 105 | |
| GenBank mRNA | BC158203.1 | - | 1 - 2 | 344 | 105 | |
More Information
Set Membership: |
|

hdhd5
Genomic Map