Assay Details
| Gene Symbol: kcnq2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114907.1 | NP_001108379.1 | 1 - 2 | 317 | 61 | |
| GenBank mRNA | BC154025.1 | - | 1 - 2 | 317 | 61 | |
More Information
Set Membership: |
|

kcnq2a
Genomic Map