Assay Details
| Gene Symbol: cxcl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115055.1 | NP_001108527.1 | 1 - 2 | 208 | 66 | |
| GenBank mRNA | AB331771.1 | - | 1 - 2 | 208 | 66 | |
More Information
Set Membership: |
|

cxcl20
Genomic Map