Assay Details
| Gene Symbol: cxcl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115055.1 | NP_001108527.1 | 2 - 3 | 336 | 85 | |
| XM_009301661.3 | XP_009299936.1 | 2 - 3 | 381 | 85 | ||
| GenBank mRNA | AB331771.1 | - | 2 - 3 | 336 | 85 | |
More Information
Set Membership: |
|

cxcl20
Genomic Map