Assay Details
| Gene Symbol: cxcl18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115060.1 | NP_001108532.1 | 2 - 3 | 235 | 74 | |
| GenBank mRNA | AB331773.1 | - | 2 - 3 | 235 | 74 | |
More Information
Set Membership: |
|

cxcl18b
Genomic Map