Assay Details
| Gene Symbol: edar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115064.2 | NP_001108536.2 | 9 - 10 | 928 | 71 | |
| GenBank mRNA | EF137867.1 | - | 9 - 10 | 928 | 71 | |
More Information
Set Membership: |
|

edar
Genomic Map