Assay Details
| Gene Symbol: stk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115092.3 | 1 - 2 | 295 | 91 | ||
| XM_003198178.5 | XP_003198226.1 | 1 - 2 | 284 | 91 | ||
| GenBank mRNA | EH542988.1 | - | 1 - 2 | 333 | 91 | |
| EH564126.1 | - | 5 - 6 | 800 | 91 | ||
| EH570280.1 | - | 5 - 6 | 826 | 91 | ||
More Information
Set Membership: |
|

stk19
Genomic Map