Assay Details
| Gene Symbol: ccl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115102.1 | NP_001108574.1 | 1 - 2 | 71 | 61 | |
| GenBank mRNA | AB331768.1 | - | 1 - 2 | 71 | 61 | |
More Information
Set Membership: |
|

ccl44
Genomic Map