Assay Details
| Gene Symbol: ccl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115102.1 | NP_001108574.1 | 2 - 3 | 204 | 59 | |
| XM_009305799.3 | XP_009304074.1 | 2 - 3 | 282 | 59 | ||
| GenBank mRNA | AB331768.1 | - | 2 - 3 | 204 | 59 | |
More Information
Set Membership: |
|

ccl44
Genomic Map