Assay Details
| Gene Symbol: dpp6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115122.1 | NP_001108594.1 | 28 - 29 | 2801 | 57 | |
| XM_005163289.3 | XP_005163346.1 | 23 - 24 | 2769 | 57 | ||
| XM_005163290.4 | XP_005163347.1 | 23 - 24 | 2244 | 57 | ||
| XM_005163291.4 | XP_005163348.1 | 23 - 24 | 2212 | 57 | ||
More Information
Set Membership: |
|

dpp6b
Genomic Map