Assay Details
| Gene Symbol: dpp6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115122.1 | NP_001108594.1 | 7 - 8 | 840 | 70 | |
| XM_005163289.3 | XP_005163346.1 | 2 - 3 | 790 | 70 | ||
| XM_005163290.4 | XP_005163347.1 | 2 - 3 | 265 | 70 | ||
| XM_005163291.4 | XP_005163348.1 | 2 - 3 | 233 | 70 | ||
More Information
Set Membership: |
|

dpp6b
Genomic Map