Assay Details
| Gene Symbol: ewsr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115138.2 | NP_001108610.1 | 13 - 14 | 1542 | 60 | |
| XM_021479109.1 | XP_021334784.1 | 13 - 14 | 1541 | 60 | ||
| GenBank mRNA | BC056281.1 | - | 13 - 14 | 1519 | 60 | |
| BC068397.1 | - | 12 - 13 | 1511 | 60 | ||
| DT075886.1 | - | 3 - 4 | 321 | 60 | ||
More Information
Set Membership: |
|

ewsr1a
Genomic Map