Assay Details
| Gene Symbol: ewsr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115138.2 | NP_001108610.1 | 2 - 3 | 82 | 77 | |
| XM_021479109.1 | XP_021334784.1 | 2 - 3 | 168 | 77 | ||
| XM_021479110.1 | XP_021334785.1 | 2 - 3 | 172 | 77 | ||
| GenBank mRNA | FP154885.1 | - | 2 - 3 | 118 | 77 | |
More Information
Set Membership: |
|

ewsr1a
Genomic Map