Assay Details
| Gene Symbol: clasp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115139.1 | NP_001108611.1 | 22 - 23 | 2306 | 103 | |
| XM_021478703.1 | XP_021334378.1 | 24 - 25 | 2868 | 103 | ||
| XM_021478705.1 | XP_021334380.1 | 24 - 25 | 2868 | 103 | ||
| XM_021478707.1 | XP_021334382.1 | 24 - 25 | 2814 | 103 | ||
| XM_021478711.1 | XP_021334386.1 | 23 - 24 | 2789 | 103 | ||
More Information
Set Membership: |
|

clasp1a
Genomic Map