Assay Details
| Gene Symbol: clasp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115139.1 | NP_001108611.1 | 30 - 31 | 3240 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

clasp1a| Gene Symbol: clasp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115139.1 | NP_001108611.1 | 30 - 31 | 3240 | 77 | |
Set Membership: |
|