Assay Details
| Gene Symbol: ppp4r2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115140.1 | NP_001108612.1 | 5 - 6 | 549 | 80 | |
| GenBank mRNA | BC060912.1 | - | 5 - 6 | 487 | 80 | |
| BC151885.1 | - | 5 - 6 | 508 | 80 | ||
More Information
Set Membership: |
|

ppp4r2a
Genomic Map