Assay Details
| Gene Symbol: ppp4r2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115140.1 | NP_001108612.1 | 8 - 9 | 1042 | 117 | |
| GenBank mRNA | BC060912.1 | - | 8 - 9 | 980 | 117 | |
| BC151885.1 | - | 8 - 9 | 1001 | 117 | ||
More Information
Set Membership: |
|

ppp4r2a
Genomic Map