Assay Details
| Gene Symbol: prlh2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115147.1 | NP_001108619.1 | 1 | 1049 | 64 | |
| GenBank mRNA | EU165206.1 | - | 1 | 1049 | 64 | |
More Information
Set Membership: |
|

prlh2r
Genomic Map