Assay Details
| Gene Symbol: cars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001118900.1 | NP_001112372.1 | 10 - 11 | 1212 | 102 | |
| XM_005168859.4 | XP_005168916.1 | 9 - 10 | 1038 | 102 | ||
| GenBank mRNA | BC048039.1 | - | 9 - 10 | 1034 | 102 | |
| BC163881.1 | - | 10 - 11 | 1178 | 102 | ||
More Information
Set Membership: |
|

cars1
Genomic Map