Assay Details
| Gene Symbol: cars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001118900.1 | NP_001112372.1 | 15 - 16 | 1786 | 66 | |
| XM_005168859.4 | XP_005168916.1 | 14 - 15 | 1612 | 66 | ||
| GenBank mRNA | BC048039.1 | - | 14 - 15 | 1608 | 66 | |
More Information
Set Membership: |
|

cars1
Genomic Map