Assay Details
| Gene Symbol: cars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001118900.1 | NP_001112372.1 | 19 - 20 | 2202 | 82 | |
| XM_005168859.4 | XP_005168916.1 | 18 - 19 | 2028 | 82 | ||
More Information
Set Membership: |
|

cars1
Genomic Map