Assay Details
| Gene Symbol: cars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001118900.1 | NP_001112372.1 | 22 - 23 | 2420 | 63 | |
| XM_005168859.4 | XP_005168916.1 | 21 - 22 | 2246 | 63 | ||
| GenBank mRNA | BC163881.1 | - | 16 - 17 | 1746 | 63 | |
More Information
Set Membership: |
|

cars1
Genomic Map