Assay Details
| Gene Symbol: cars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001118900.1 | NP_001112372.1 | 6 - 7 | 705 | 77 | |
| XM_005168859.4 | XP_005168916.1 | 5 - 6 | 531 | 77 | ||
| GenBank mRNA | BC163881.1 | - | 6 - 7 | 671 | 77 | |
More Information
Set Membership: |
|

cars1
Genomic Map