Assay Details
| Gene Symbol: magi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327853.1 | NP_001314782.1 | 20 - 21 | 3848 | 69 | |
| XM_017355040.2 | XP_017210529.1 | 20 - 21 | 3889 | 69 | ||
| XM_017355041.2 | XP_017210530.1 | 20 - 21 | 3886 | 69 | ||
| XM_017355042.2 | XP_017210531.1 | 20 - 21 | 3889 | 69 | ||
| XM_017355043.2 | XP_017210532.1 | 18 - 19 | 3683 | 69 | ||
| XM_017355044.2 | XP_017210533.1 | 18 - 19 | 3681 | 69 | ||
| XM_017355045.2 | XP_017210534.1 | 20 - 21 | 3895 | 69 | ||
| XM_017355046.2 | XP_017210535.1 | 20 - 21 | 3894 | 69 | ||
| XM_017355047.2 | XP_017210536.1 | 20 - 21 | 3895 | 69 | ||
| XM_017355048.2 | XP_017210537.1 | 20 - 21 | 3895 | 69 | ||
| XM_021475052.1 | XP_021330727.1 | 20 - 21 | 3885 | 69 | ||
More Information
Set Membership: |
|

magi2a
Genomic Map