Assay Details
| Gene Symbol: magi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017355045.2 | XP_017210534.1 | 21 - 22 | 4023 | 74 | |
| XM_017355046.2 | XP_017210535.1 | 21 - 22 | 4022 | 74 | ||
More Information
Set Membership: |
|

magi2a
Genomic Map