Assay Details
| Gene Symbol: magi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017355045.2 | XP_017210534.1 | 22 - 23 | 4129 | 74 | |
| XM_017355048.2 | XP_017210537.1 | 22 - 23 | 4151 | 74 | ||
More Information
Set Membership: |
|

magi2a
Genomic Map