Assay Details
| Gene Symbol: magi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327853.1 | NP_001314782.1 | 15 - 16 | 2757 | 57 | |
| XM_017355040.2 | XP_017210529.1 | 15 - 16 | 2798 | 57 | ||
| XM_017355041.2 | XP_017210530.1 | 15 - 16 | 2795 | 57 | ||
| XM_017355042.2 | XP_017210531.1 | 15 - 16 | 2798 | 57 | ||
| XM_017355043.2 | XP_017210532.1 | 13 - 14 | 2592 | 57 | ||
| XM_017355044.2 | XP_017210533.1 | 13 - 14 | 2590 | 57 | ||
| XM_017355045.2 | XP_017210534.1 | 15 - 16 | 2804 | 57 | ||
| XM_017355046.2 | XP_017210535.1 | 15 - 16 | 2803 | 57 | ||
| XM_017355047.2 | XP_017210536.1 | 15 - 16 | 2804 | 57 | ||
| XM_017355048.2 | XP_017210537.1 | 15 - 16 | 2804 | 57 | ||
| XM_021475052.1 | XP_021330727.1 | 15 - 16 | 2800 | 57 | ||
| GenBank mRNA | AI584740.1 | - | 1 - 2 | 112 | 57 | |
More Information
Set Membership: |
|

magi2a
Genomic Map