Assay Details
| Gene Symbol: magi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327853.1 | NP_001314782.1 | 18 - 19 | 3485 | 65 | |
| XM_017355040.2 | XP_017210529.1 | 18 - 19 | 3526 | 65 | ||
| XM_017355041.2 | XP_017210530.1 | 18 - 19 | 3523 | 65 | ||
| XM_017355042.2 | XP_017210531.1 | 18 - 19 | 3526 | 65 | ||
| XM_017355043.2 | XP_017210532.1 | 16 - 17 | 3320 | 65 | ||
| XM_017355044.2 | XP_017210533.1 | 16 - 17 | 3318 | 65 | ||
| XM_017355045.2 | XP_017210534.1 | 18 - 19 | 3532 | 65 | ||
| XM_017355046.2 | XP_017210535.1 | 18 - 19 | 3531 | 65 | ||
| XM_017355047.2 | XP_017210536.1 | 18 - 19 | 3532 | 65 | ||
| XM_017355048.2 | XP_017210537.1 | 18 - 19 | 3532 | 65 | ||
More Information
Set Membership: |
|

magi2a
Genomic Map