Assay Details
| Gene Symbol: sort1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126465.1 | NP_001119937.1 | 19 - 20 | 2566 | 73 | |
| GenBank mRNA | AW184692.1 | - | 2 - 3 | 203 | 73 | |
More Information
Set Membership: |
|

sort1b
Genomic Map