Assay Details
| Gene Symbol: sort1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126465.1 | NP_001119937.1 | 1 - 2 | 360 | 108 | |
| GenBank mRNA | AW202779.1 | - | 1 - 2 | 108 | ||
| AW233291.1 | - | 1 - 2 | 351 | 108 | ||
More Information
Set Membership: |
|

sort1b
Genomic Map