Assay Details
| Gene Symbol: sort1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126465.1 | NP_001119937.1 | 9 - 10 | 1190 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sort1b| Gene Symbol: sort1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126465.1 | NP_001119937.1 | 9 - 10 | 1190 | 79 | |
Set Membership: |
|