Assay Details
| Gene Symbol: pllp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122699.1 | NP_001116171.1 | 1 - 2 | 224 | 117 | |
| XM_005159107.4 | XP_005159164.1 | 2 - 3 | 296 | 117 | ||
| GenBank mRNA | BC075971.1 | - | 1 - 2 | 225 | 117 | |
| BC146728.1 | - | 1 - 2 | 228 | 117 | ||
More Information
Set Membership: |
|

pllp
Genomic Map