Assay Details
| Gene Symbol: smad4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122700.1 | NP_001116172.1 | 2 - 3 | 654 | 84 | |
| XM_017357976.2 | XP_017213465.1 | 2 - 3 | 634 | 84 | ||
| XM_017357977.2 | XP_017213466.1 | 2 - 3 | 634 | 84 | ||
| XM_017357978.2 | XP_017213467.1 | 2 - 3 | 633 | 84 | ||
| GenBank mRNA | EU489481.1 | - | 2 - 3 | 654 | 84 | |
| EU489482.1 | - | 1 - 2 | 244 | 84 | ||
More Information
Set Membership: |
|

smad4a
Genomic Map