Assay Details
| Gene Symbol: abl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122705.1 | NP_001116177.1 | 3 - 4 | 566 | 63 | |
| XM_005166889.4 | XP_005166946.1 | 3 - 4 | 1182 | 63 | ||
| XM_005166890.4 | XP_005166947.1 | 3 - 4 | 1179 | 63 | ||
| XM_005166891.4 | XP_005166948.1 | 3 - 4 | 873 | 63 | ||
| XM_017357363.2 | XP_017212852.1 | 3 - 4 | 754 | 63 | ||
More Information
Set Membership: |
|

abl2
Genomic Map