Assay Details
| Gene Symbol: irf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122710.1 | NP_001116182.1 | 1 - 2 | 201 | 75 | |
| XM_005163652.4 | XP_005163709.1 | 2 - 3 | 930 | 75 | ||
More Information
Set Membership: |
|

irf4a
Genomic Map