Assay Details
| Gene Symbol: irf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122710.1 | NP_001116182.1 | 7 - 8 | 1248 | 76 | |
| XM_005163652.4 | XP_005163709.1 | 8 - 9 | 1977 | 76 | ||
More Information
Set Membership: |
|

irf4a
Genomic Map