Assay Details
| Gene Symbol: tafa5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122744.1 | NP_001116216.1 | 2 - 3 | 324 | 58 | |
| XM_005168534.4 | XP_005168591.1 | 3 - 4 | 1186 | 58 | ||
| XM_005168535.4 | XP_005168592.1 | 3 - 4 | 1059 | 58 | ||
More Information
Set Membership: |
|

tafa5a
Genomic Map