Assay Details
| Gene Symbol: dgcr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122749.1 | NP_001116221.1 | 7 - 8 | 2055 | 63 | |
| GenBank mRNA | BC124419.1 | - | 6 - 7 | 1022 | 63 | |
More Information
Set Membership: |
|

dgcr8
Genomic Map