Assay Details
| Gene Symbol: matr3l1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122784.2 | NP_001116256.2 | 12 - 13 | 2513 | 72 | |
| XM_005173122.4 | XP_005173179.1 | 12 - 13 | 2523 | 72 | ||
| XM_005173123.4 | XP_005173180.1 | 12 - 13 | 2523 | 72 | ||
| XM_009291035.3 | XP_009289310.1 | 11 - 12 | 1622 | 72 | ||
| GenBank mRNA | CK687062.1 | - | 2 - 3 | 186 | 72 | |
More Information
Set Membership: |
|

matr3l1.2
Genomic Map