Assay Details
| Gene Symbol: matr3l1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005173123.4 | XP_005173180.1 | 15 - 16 | 2792 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

matr3l1.2| Gene Symbol: matr3l1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005173123.4 | XP_005173180.1 | 15 - 16 | 2792 | 114 | |
Set Membership: |
|