Assay Details
| Gene Symbol: hmgb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122836.1 | NP_001116308.1 | 4 - 5 | 585 | 62 | |
| GenBank mRNA | BC139583.1 | - | 4 - 5 | 553 | 62 | |
More Information
Set Membership: |
|

hmgb3a
Genomic Map