Assay Details
| Gene Symbol: colec12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122840.1 | NP_001116312.1 | 6 - 7 | 2251 | 55 | |
| XM_005163457.4 | XP_005163514.1 | 6 - 7 | 2281 | 55 | ||
More Information
Set Membership: |
|

colec12
Genomic Map