Assay Details
| Gene Symbol: dpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122846.2 | NP_001116318.1 | 3 - 4 | 252 | 113 | |
| GenBank mRNA | BC093353.1 | - | 3 - 4 | 233 | 113 | |
| EH452076.1 | - | 3 - 4 | 228 | 113 | ||
More Information
Set Membership: |
|

dpm2
Genomic Map