Assay Details
| Gene Symbol: acyp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122849.1 | NP_001116321.1 | 2 - 3 | 179 | 98 | |
| GenBank mRNA | EB939010.1 | - | 2 - 3 | 179 | 98 | |
More Information
Set Membership: |
|

acyp1
Genomic Map