Assay Details
| Gene Symbol: dab2ipb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122857.2 | NP_001116329.1 | 2 - 3 | 569 | 59 | |
| XM_005171880.4 | XP_005171937.1 | 2 - 3 | 564 | 59 | ||
| XM_005171881.4 | XP_005171938.1 | 2 - 3 | 295 | 59 | ||
| XM_009301511.3 | XP_009299786.1 | 2 - 3 | 170 | 59 | ||
More Information
Set Membership: |
|

dab2ipb
Genomic Map