Assay Details
| Gene Symbol: hsbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200754.2 | NP_957048.1 | 3 - 4 | 365 | 105 | |
| GenBank mRNA | BC059566.1 | - | 3 - 4 | 325 | 105 | |
| EH450852.1 | - | 3 - 4 | 387 | 105 | ||
More Information
Set Membership: |
|

hsbp1b
Genomic Map