Assay Details
| Gene Symbol: derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200903.2 | NP_957197.2 | 1 - 2 | 307 | 116 | |
| GenBank mRNA | BC064666.1 | - | 1 - 2 | 306 | 116 | |
| CK026866.1 | - | 1 - 2 | 233 | 116 | ||
More Information
Set Membership: |
|

derl3
Genomic Map