Assay Details
| Gene Symbol: nt5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200932.1 | NP_957226.1 | 2 - 3 | 576 | 62 | |
| GenBank mRNA | BC055243.1 | - | 2 - 3 | 576 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nt5e| Gene Symbol: nt5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200932.1 | NP_957226.1 | 2 - 3 | 576 | 62 | |
| GenBank mRNA | BC055243.1 | - | 2 - 3 | 576 | 62 | |
Set Membership: |
|