Assay Details
| Gene Symbol: cpeb4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351720.1 | NP_001338649.1 | 3 - 4 | 1340 | 119 | |
| NM_200981.2 | NP_957275.2 | 3 - 4 | 1292 | 119 | ||
| XM_017359000.2 | XP_017214489.1 | 3 - 4 | 1368 | 119 | ||
| GenBank mRNA | BC055522.1 | - | 3 - 4 | 1292 | 119 | |
| BC165168.1 | - | 2 - 3 | 1035 | 119 | ||
| EF033337.1 | - | 2 - 3 | 1487 | 119 | ||
More Information
Set Membership: |
|

cpeb4a
Genomic Map